Genistein
Glass bottle for solids, 1 g
Catalogus # CAYM10005167-1
Leverancier: Cayman Chemical
Ga naar:
Product Details & Documents
Synoniemen: 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one, 4',6'-Dihydroxy-3-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',5,7-Trihydroxyisoflavone
- CAS-nummer:446-72-0
- Formule:C₁₅H₁₀O₅
- MDL-nummer:MFCD00016952
- Molecular Weight:270.24 g/mol
- Smelten Pt:297…298 °C
- EINECS:207-174-9
Documents
Certificaten
Zoeken op lotnummerSpecificaties
Specificaties
- Verpakking:Glass bottle for solids
- VPE:1 g
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



