3,4',5-Trimethoxybenzophenone
Glass bottle for solids, 25 mg
Catalog # CAYM10004185-25
Supplier: Cayman Chemical
3,4',5-Trimethoxybenzophenone
Catalog # CAYM10004185-25
Supplier: Cayman Chemical
CAS Number: 94709-12-3
Jump to:
Product Details & Documents
- CAS Number:94709-12-3
- Formula:C₁₆H₁₆O₄
Specifications
Specifications
- Verpakking:Glass bottle for solids
- VPE:25 mg



