Multi-way stopcocks, Discofix®
Three-way stopcocks with 500 mm extension
Catalogus # BRAU4098501
Leverancier: B. Braun
Ga naar:
Product Details & Documents
Suitable for all pressure infusion systems <2 bar.
- Continuous flow channels for accurate pressure monitoring
- Swivel lock can be moved axially or radially for fast and secure connection
- PVC and latex-free
Suitable for all pressure infusion systems <2 bar.
Documents
Specificaties
- Omschrijving:Three-way stopcocks with 500 mm extension