Weighing pipettes
10 ml
Catalogus # 614-0153
Leverancier: GERBER FUNKE
Weighing pipettes
10 ml
Ga naar:
Product Details & Documents
Curved
Specificaties
- Capaciteit:10 ml
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
#: 20864.320
Leverancier: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83640.320
Leverancier: VWR Chemicals
Acetonitril ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
#: 1.00030.2500
Leverancier: Merck
Natriumhydroxide 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur. volumetric solution
#: 31770.294
Leverancier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83645.320
Leverancier: VWR Chemicals
Bufferoplossingen, gebruiksklaar, waarnaar wordt verwezen bij 20 °C, CertiPUR®, Supelco®
#: 1.09439.1000
Leverancier: Merck



