Orders LinkContactUsLinkComponent
Milk butyrometer
Butyrometer for milk, 0 - 9% : 0,1
Catalogus # 614-0093
Leverancier:   GERBER FUNKE
Milk butyrometer
Catalogus # 614-0093
Leverancier:  GERBER FUNKE
Milk butyrometer
Milk butyrometer
Butyrometer for milk, 0 - 9% : 0,1
Price
122,00 €
Log in voor beschikbaarheid
1 * 10 ST
Product Details & Documents
Borosilicate glass.
  • With or without stopper
Available with a declaration of conformity per batch and type (in German only).
Specificaties
  • Omschrijving:
    Butyrometer for milk, 0 - 9% : 0,1
  • Seq:
    07
Accessories
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Stopper, rubber, conical, for all measuring method butyrometers
    Stopper, rubber, conical, for all measuring method butyrometers
    614-2028
    Log in voor beschikbaarheid
    1 * 100 ST - 48,20 €