Digital stereo microscope, DM-143-FBGG-C
Reticle with 100 divisions in 10 mm (Ø = 23 mm)
630-2784
: MOTIC
This microscope features a binocular head, 45º inclined, with built-in digital camera (3 MP). It can be connected to a computer via USB port for viewing and capturing live images.
- Greenough zoom optical system
- Coarse focusing system with tension adjustment
- Incident and transmitted halogen illumination with intensity control
- Pole type stand with large working base
- Large dual focusing knobs with slip clutch to prevent damage to mechanism
System requirements:
Windows XP or above, MAC OSX
Intel Pentium
1 Ghz processor, video card: 128 MB
RAM 512 MB or above
CD-ROM drive
USB 2.0
: Supplied with software Motic Images Plus 2.0 for PC & MAC, dust cover and USB cable.
- Omschrijving:Reticle with 100 divisions in 10 mm (Ø = 23 mm)
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.