Support clamps
DN 150, Schott "Kugel-Flanges" (KF)
Catalogus # BOCH17216
Leverancier: BOCHEM
Ga naar:
Product Details & Documents
Support clamps made of stainless steel (18/19) and PTFE for either Schott laboratory flat flanges (PF) (optimal flange angle 45°) or Schott "Kugel-Flanges" (ball and socket system) (KF).
- For reactors and other vessels
- User friendly thanks to joint screws
- With two supporting devices for 26,9 mm tubes
Specificaties
- Voor:Schott "Kugel-Flanges" (KF)
- Omschrijving:DN 150