Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Splash heads, with sinter, Quickfit®
Splash heads, with sinter, Quickfit®
29/32 NS, 29/32 NS
Catalogus # 201-0576
Leverancier:   SciLabware
Splash heads, with sinter, Quickfit®
Catalogus # 201-0576
Leverancier:  SciLabware
Splash heads, with sinter, Quickfit®
Splash heads, with sinter, Quickfit®
29/32 NS, 29/32 NS
Price
181,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
PYREX® borosilicate glas
  • Prevents possible contamination of the condenser from 'bumping' solvents
  • Features a coarse porosity 1 sinter to reduce carry over
  • Splash heads feature a 29/32 socket
For use with rotary evaporators.
Specificaties
  • Afmetingen kern:
    29/32 NS
  • Huls:
    29/32 NS
  • Seq:
    03
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.