Brushes, FlexBrush™
Polyester cover, NovaPoly
Catalogus # 141-1016
Leverancier: MICRONOVA
Ga naar:
Product Details & Documents
The FlexBrush™ is ideal for cleaning over and around piping and fittings. The flexibility of the head allows the user to shape the head for custom cleaning applications.
- Bendable brush with polyester or microfibre cover
- Acme threaded end attaches to a variety of handles for extended reach
- Threaded handle (STA series) can be added for extended reach
- Covers are compatible with strong disinfectants and autoclavable
Specificaties
- Omschrijving:Polyester cover, NovaPoly
- Seq:05
- Cat. No.:141-1016
- Environmentally Preferable:Afvalvermindering