Spanner wrenches
Spanner wrench, for nuts Ø 58 - 62 mm, for HiScale™ 16 columns, Gegalvaniseerd Staal
Catalogus # 28-9647-76
Leverancier: Cytiva
Ga naar:
Product Details & Documents
These spanner wrenches are used with HiScale™ columns. To readjust the position of the adapter in the column a spanner can be used to turn the end cap. To prevent the column from rotating, a second spanner can be used to hold the end housing.
Specificaties
- Omschrijving:Spanner wrench, for nuts Ø 58 - 62 mm, for HiScale™ 16 columns
- Materiaal:Gegalvaniseerd Staal
- Seq:01