Safety spectacles, 511
Anti-UV, anti-scratch, anti-fog, zwart, Clear PC
Catalog # UNVT511.00.03.01
Supplier: UNIVET
Jump to:
Product Details & Documents
Safety spectacles, 511 has an essential design which offers good performance and the possibility of many different lenses, to meet different needs.
- Adjustment of temples in length and inclination
- Built-in side shields
- Frame made of an allergic material
- Mirrored version
These spectacles are used for mechanical works in good visibility conditions.
Wearing these spectacles for prolonged period of time will not create distortion or fatigue.
Weight: 34 g
Specifications
- Type:Anti-UV, anti-scratch, anti-fog
- Kleur:zwart
- Seq:02
- Beglazing:Clear PC