Petrischalen
Met drie ventilatieopeningen, 90 mm, 15 mm, Aseptisch, 25 zakken met 20 stuks
Leverancier: Anicrin
Ga naar:
Product Details & Documents
PS. Suitable for all microbiology applications.
Specificaties
- Ø:90 mm
- Verpakt:25 zakken met 20 stuks
- Steriliteit:Aseptisch
- Uitvoering:Met drie ventilatieopeningen
- Hoogte:15 mm
- Seq:05
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



