Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

Petrischalen

With 3 vents, 60 mm, 8 mm, Aseptisch, 50×10/bag
  ANIC05.80.18PADV
 :   Anicrin
Petrischalen
  ANIC05.80.18PADV
 :  Anicrin
Petrischalen
Petrischalen

With 3 vents, 60 mm, 8 mm, Aseptisch, 50×10/bag

 
116,00 €
   
1 * 500 ST
 
  • Ø:
    60 mm
  • Verpakt:
    50×10/bag
  • Steriliteit:
    Aseptisch
  • Uitvoering:
    With 3 vents
  • Hoogte:
    8 mm
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.