Orders LinkContactUsLinkComponent
Cryoboxen, karton
Cryoboxen, karton
-, 4×4, 30 mm
Catalog # HERA820016
Supplier:   Thermo Scientific
Cryoboxen, karton
Catalog # HERA820016
Supplier:  Thermo Scientific
Cryoboxen, karton
Cryoboxen, karton
-, 4×4, 30 mm
Price
4,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
Cryobestendig karton.
  • Use with cardboard storage boxes to divide box into grids for vial storage
  • Box dividers for cold storage application available in 7×7 , 8×8, 9×9 or 10×10 grids
Voor opslag van cryovials bij lage temperaturen.
Documents
Specifications
  • Compartimenten:
    4×4
  • Voor buizen Ø:
    30 mm
  • Omschrijving:
    -
  • Seq:
    05