Cryoboxen, karton
-, 4×4, 30 mm
Catalog # HERA820016
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
Cryobestendig karton.
- Use with cardboard storage boxes to divide box into grids for vial storage
- Box dividers for cold storage application available in 7×7 , 8×8, 9×9 or 10×10 grids
Voor opslag van cryovials bij lage temperaturen.
Documents
HERA820002_TDS_EN
Datasheet
Specifications
- Compartimenten:4×4
- Voor buizen Ø:30 mm
- Omschrijving:-
- Seq:05
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.