Labels, hazard warning
Hazard labels: laser beam, PIC 309-TRI 315-B7541, Polyester, zwart op geel, 315×273 mm, 1 label/card
Catalogus # BRDY223395
Leverancier: Brady
Ga naar:
Product Details & Documents
These self adhesive signs are designed to warn people to be careful and to take precautions of potential hazards.
Available in different sizes.
Certificeringen: Conforms to Health and Safety (Safety Signs and Signals) Regulations 1996.
Documents
Specificaties
- Kleur:zwart op geel
- Omschrijving:Hazard labels: laser beam
- Afmetingen:315×273 mm
- Materiaal:Polyester
- Seq:45
- Model:PIC 309-TRI 315-B7541
- Verpakt:1 label/card
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



