Labels, hazard warning
Hazard labels: danger asbestos, —, Polyester, zwart op geel, 26×52 mm, 250 labels/roll
Catalogus # BRDY223337
Leverancier: Brady
Ga naar:
Product Details & Documents
These self adhesive signs are designed to warn people to be careful and to take precautions of potential hazards.
Available in different sizes.
Certifications: Conforms to Health and Safety (Safety Signs and Signals) Regulations 1996.
Specificaties
- Kleur:zwart op geel
- Omschrijving:Hazard labels: danger asbestos
- Afmetingen:26×52 mm
- Materiaal:Polyester
- Seq:08
- Model:—
- Verpakt:250 labels/roll
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



