Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
[EN]MEPS -C2-XCHANGE 23GA BIN PK5 1 * 5 ST
549-0751
:
Trajan Scientific and Medical
[EN]MEPS -C2-XCHANGE 23GA BIN PK5 1 * 5 ST
549-0751
:
Trajan Scientific and Medical
SGE Syringes for the Laboratory
Trajan Scientific and Medical GC Consumables
Trajan Scientific Syringes for the Laboratory
Frequently Bought Together
Aceton ≥99%, TECHNICAL
: VWR Chemicals
Sealing film for manual application, Parafilm® M
: HEATHROW
VWR®, Nitrilhandschoenen
: VWR Collection
VWR®, gezichtstissues
: VWR Collection
pH-teststrips, MQuant®, Supelco®
: Merck
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR®, Transferpipetten, Hoge Prestaties
: VWR Collection
Acetonitril, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrilhandschoenen
: VWR Collection
Acetonitril ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Nitril handschoenen, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Serologische pipetten, standaardreeks
: VWR Collection