Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Blood collection needles, BD Vacutainer® Eclipse™
Blood collection needles, BD Vacutainer® Eclipse™
22, 32 mm, zwart
Catalogus # BDAM368651
Leverancier:   BD Medical
Blood collection needles, BD Vacutainer® Eclipse™
Catalogus # BDAM368651
Leverancier:  BD Medical
Blood collection needles, BD Vacutainer® Eclipse™
Blood collection needles, BD Vacutainer® Eclipse™
22, 32 mm, zwart
Price
71,00 €
Log in voor beschikbaarheid
1 * 100 ST
Product Details & Documents
The BD Vacutainer® Eclipse™ blood collection needle is a safety-engineered, multi-sample blood collection needle that offers a simple, effective way to collect blood while reducing the possibility of needlestick injuries. It features a safety shield that allows for one-handed activation to cover the needle immediately upon withdrawal from the vein and confirms proper activation with an audible click.
  • Built-in safety shield is aligned with the bevel of the needle
  • The safety shield permanently locks into place, significantly reducing the risk of needlestick injuries
  • The pre-attached holder (only supplied with some models) minimises the risk of accidental needlestick injuries from the holder end of the device
Specificaties
  • Kleurcode:
    zwart
  • Meter:
    22
  • Lengte:
    32 mm
  • Seq:
    04
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.