Extraction tubes, Mojonnier
Extraction tubes, Mojonnier, with flattened bulb
Catalogus # 614-0187
Leverancier: GERBER FUNKE
Ga naar:
Product Details & Documents
According to Mojonnier`s method complete with cork stopper.
Documents
GERB3871_TDS_EN
Technische fiche
Specificaties
- Omschrijving:Extraction tubes, Mojonnier, with flattened bulb
Accessories
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Cork stopper for extraction tubes, Mojonnier`s methodCork stopper for extraction tubes, Mojonnier`s method614-0188Log in voor beschikbaarheid1 * 1 ST - 0,40 €
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Stand, wooden, for 12 extraction tubes acc. to Mojonnier`s methodStand, wooden, for 12 extraction tubes acc. to Mojonnier`s method614-0189Log in voor beschikbaarheid1 * 1 ST - 86,90 €