Labels, hazard warning
Hazard labels: handling vehicle, —, Polyester, zwart op geel, 200×173 mm, 1 label/card
Catalogus # BRDY250220
Leverancier: Brady
Ga naar:
Product Details & Documents
These self adhesive signs are designed to warn people to be careful and to take precautions of potential hazards.
Available in different sizes.
Certifications: Conforms to Health and Safety (Safety Signs and Signals) Regulations 1996.
Specificaties
- Kleur:zwart op geel
- Omschrijving:Hazard labels: handling vehicle
- Afmetingen:200×173 mm
- Materiaal:Polyester
- Seq:35
- Model:—
- Verpakt:1 label/card