Orders LinkContactUsLinkComponent
Gastro-norm container
Gastro-norm container
Catalogus # HUPF0131642H
Leverancier:   HUPFER METALLWERKE
Gastro-norm container
Catalogus # HUPF0131642H
Leverancier:  HUPFER METALLWERKE
Gastro-norm container
Gastro-norm container
Price
67,10 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Stainless steel 18/10.
  • Fast, easy cleaning
  • Good chemical resistance
  • Very good thermal resistance
Multipurpose containers for a wide range of uses.