Razor blades, Gem®
Eenzijdige scheermesjes, Gem® Uncoated, 3 facetten, ontvette roestvrijstalen achterzijde, 100 mesjes per dispenser, Roestvrij staal, 0,22 mm, 38×19 mm
Catalog # PERSAGBL-7029-0000
Supplier: PERSONNA MEDICAL
Jump to:
Product Details & Documents
Razor blades for catheter cutting or cleanroom applications.
- High precision, sharpness and safety
- Made from carbon steel or from stainless steel
Carbon steel blades are suitable for ISO Class 7 cleanroom environments.
Stainless steel blades are suitable for ISO Class 5, 6 and 7 cleanroom environments.
Specifications
- Omschrijving:Eenzijdige scheermesjes, Gem® Uncoated, 3 facetten, ontvette roestvrijstalen achterzijde, 100 mesjes per dispenser
- l×b:38×19 mm
- Materiaal:Roestvrij staal
- Dikte:0,22 mm