Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Recombinant streptavidin

Recombinant streptavidin

Streptavidin, recombinant, 100 mg
Catalogus # 786-586
Leverancier:   G-Biosciences
Recombinant streptavidin
Catalogus # 786-586
Leverancier:  G-Biosciences
CAS-nummer:  
Recombinant streptavidin
Recombinant streptavidin

Streptavidin, recombinant, 100 mg

CAS-nummer:  
Price
1 940,00 €
Log in voor beschikbaarheid
1 * 100 mg
Product Details & Documents

Recombinant streptavidin from Streptomyces avidinii, produced in E.coli.

Streptavidin is a tetrameric protein that has no carbohydrate and thereby reduces the amount of non specific binding. The solubility of streptavidin (isoelectric pH 5) in aqueous buffer is lower than avidin, but the binding of streptavidin to biotin is similar to that of avidin.

Documents

Specificaties

Specificaties

  • VPE:
    100 mg
  • Omschrijving:
    Streptavidin, recombinant

Specificaties