Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Selenium dioxide ≥99.4% (metals basis)

Selenium dioxide ≥99.4% (metals basis)

Glass bottle for solids, 1 kg
Catalogus # 12358.A1
Giftig
Gezondheid
Milieu
Leverancier:   Thermo Fisher Scientific
Selenium dioxide ≥99.4% (metals basis)
Catalogus # 12358.A1
CAS-nummer:  7446-08-4
Selenium dioxide ≥99.4% (metals basis)
Selenium dioxide ≥99.4% (metals basis)

Glass bottle for solids, 1 kg

CAS-nummer:  7446-08-4
Price
Informeer naar de prijs
Log in voor beschikbaarheid
1 * 1 kg
Product Details & Documents

Purity: 99.4% CAS No.: 7446-08-4 Molecular Formula: O2Se Formula Weight: 110.96 Physical Form: Powder Density: 3.95 Appearance: White to pink Odour: Odourless Melting Point: 340-350° subl. Solubility: Partially soluble in water, methanol, ethanol, acetone. Soluble in concentrated H2SO4 Merck Reference: 14,8434 UN No.: 3283 MDL No.: MFCD00003562

As an oxidizing agent, in manufacturing of other selenium compounds

  • CAS-nummer:7446-08-4
  • Formule:SeO₂
  • MDL-nummer:MFCD00003562
  • Molecular Weight:110.96 g/mol
  • Merck index:14,8434
  • Smelten Pt:340…350 °C (sublimatie)
  • Dichtheid:3,95
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:231-194-7
  • UN:3283
  • ADR:6.1,II
Specificaties

Specificaties

  • Verpakking:
    Glass bottle for solids
  • VPE:
    1 kg

Specificaties