Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Xyleen, extra pure

Xyleen, extra pure

Plastic bottle, 10 L
Catalogus # KRAF07048.6700
Ontvlambaar
Irriterend
Leverancier:   Bernd Kraft
Xyleen, extra pure
Catalogus # KRAF07048.6700
Leverancier:  Bernd Kraft
CAS-nummer:  1330-20-7
Xyleen, extra pure
Xyleen, extra pure

Plastic bottle, 10 L

CAS-nummer:  1330-20-7
Price
183,00 €
Log in voor beschikbaarheid
1 * 10 L
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
Synoniemen:  Dimethylbenzene
  • CAS-nummer:1330-20-7
  • Formule:C₆H₄(CH₃)₂
  • MDL-nummer:MFCD00077264
  • Molecular Weight:106.17 g/mol
  • Merck index:13,10136
  • Kokend Pt:140 °C (1013 hPa)
  • Smelten Pt:–34 °C
  • Dichtheid:0,86…0.88 g/cm³ (20 °C)
  • Flits Pt:25 °C
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:215-535-7
  • UN:1307
  • ADR:3,III
Specificaties

Specificaties

  • Verpakking:
    Plastic bottle
  • VPE:
    10 L

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.