Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Natriumhydroxide 10% in aqueous solution, extra pure

Natriumhydroxide 10% in aqueous solution, extra pure

Plastic bottle, 20 L
Catalogus # KRAF05093.7020
Corrosief
Leverancier:   Bernd Kraft
Natriumhydroxide 10% in aqueous solution, extra pure
Catalogus # KRAF05093.7020
Leverancier:  Bernd Kraft
CAS-nummer:  1310-73-2
Natriumhydroxide 10% in aqueous solution, extra pure

Plastic bottle, 20 L

CAS-nummer:  1310-73-2
Price
229,00 €
Log in voor beschikbaarheid
1 * 20 L
Product Details & Documents
Synoniemen:  Caustic soda
  • CAS-nummer:1310-73-2
  • Formule:NaOH
  • MDL-nummer:MFCD00003548
  • Molecular Weight:40 g/mol
  • Kokend Pt:119…135 °C (1013 hPa)
  • Smelten Pt:9…10 °C
  • Dichtheid:1,53 g/cm³ (20 °C)
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:215-185-5
  • UN:1824
  • ADR:8,II

Documents

Specificaties

Specificaties

  • Verpakking:
    Plastic bottle
  • VPE:
    20 L

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.