Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Natriumhydroxide 10% in aqueous solution, extra pure

Natriumhydroxide 10% in aqueous solution, extra pure

Plastic bottle, 1 L
Catalogus # KRAF05093.3000
Corrosief
Leverancier:   Bernd Kraft
Natriumhydroxide 10% in aqueous solution, extra pure
Catalogus # KRAF05093.3000
Leverancier:  Bernd Kraft
CAS-nummer:  1310-73-2
Natriumhydroxide 10% in aqueous solution, extra pure

Plastic bottle, 1 L

CAS-nummer:  1310-73-2
Price
30,60 €
Log in voor beschikbaarheid
1 * 1 L
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
Synoniemen:  Caustic soda
  • CAS-nummer:1310-73-2
  • Formule:NaOH
  • MDL-nummer:MFCD00003548
  • Molecular Weight:40 g/mol
  • Kokend Pt:119…135 °C (1013 hPa)
  • Smelten Pt:9…10 °C
  • Dichtheid:1,53 g/cm³ (20 °C)
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:215-185-5
  • UN:1824
  • ADR:8,II

Documents

Specificaties

Specificaties

  • Verpakking:
    Plastic bottle
  • VPE:
    1 L

Specificaties