Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

Diallylamine

15 kg
  8.03521.9015
 
 
 
 :   Merck
Diallylamine
  8.03521.9015
 :  Merck
 :  124-02-7
Diallylamine

15 kg

 :  124-02-7
 
 
   
1 * 15 kg
 
  •  :124-02-7
  •  :C₆H₁₁N
  •  :MFCD00008642
  •  :97.16 g/mol
  •  :Kamertemperatuur
  •  :3 (6.1, 8),II
  •  :M-Clarity™ quality level = MQ200
  •  :204-671-2
  •  :2359

Documents

 

 

  • Solubility:
    86 g/l (20 °C)
  • Melting Point:
    -88 °C
  • Molar Mass:
    97.15 g/mol
  • Boiling Point:
    111 °C (1013 hPa)
  • Vapor Pressure:
    24.2 hPa (20 °C)
  • Flash Point:
    11 °C
  • Refractive Index:
    1.4405 (20 °C, 589 nm)
  • Explosion Limit:
    0.93 - 14.2 %(V)
  • Density:
    0.79 g/cm3 (20 °C)
  • pH:
    11.5 (9.7 g/l, H2O)
  • Ignition Point:
    273 °C

 

  • VPE:
    15 kg