GC septa
EC septa for longer injection life, low bleed and low injection port adhesion, 11,5 mm, 400 °C
Catalogus # 568-0364
Leverancier: Trajan Scientific and Medical
Ga naar:
Product Details & Documents
Specificaties
- Ø:11,5 mm
- Max. temp.:400 °C
- Omschrijving:EC septa for longer injection life, low bleed and low injection port adhesion
- Seq:14