GC septa
HT septa for optimised bleed and temperature, combined with outstanding mechanical properties, 11,5 mm, 400 °C
Catalog # 568-0363
Supplier: Trajan Scientific and Medical
Jump to:
Product Details & Documents
Specifications
- Ø:11,5 mm
- Max. temp.:400 °C
- Omschrijving:HT septa for optimised bleed and temperature, combined with outstanding mechanical properties
- Seq:13