Reaction vessels with frame assemblies
1000 ml, flens NW 100, 210 mm, DN 15
Catalogus # KGWARK-FB2
Leverancier: KGW
Ga naar:
Product Details & Documents
Reaction vessel with stationary frame, J valve and accessories
Specificaties
- Aansluiting:DN 15
- Met:flens NW 100
- Capaciteit:1000 ml
- Hoogte:210 mm
- Seq:01