Orders LinkContactUsLinkComponent
Dry buffer packs, JAW™
Dry buffer packs, JAW™
Phosphate buffered saline (PBS), JAW™, 10X, dry buffer pack for 10 l of 1X/ pack, 43 mM NaH₂PO₄, 1,37 M NaCl, 27mM KCl and 14 mM KH₂PO₄, 7,4, 2
Catalog # RC-147
Supplier:   G-Biosciences
Dry buffer packs, JAW™
Catalog # RC-147
Supplier:  G-Biosciences
CAS Number:  
Dry buffer packs, JAW™
Phosphate buffered saline (PBS), JAW™, 10X, dry buffer pack for 10 l of 1X/ pack, 43 mM NaH₂PO₄, 1,37 M NaCl, 27mM KCl and 14 mM KH₂PO₄, 7,4, 2
CAS Number:  
Price
130,00 €
Log in for availability
1 * 2 ST
Product Details & Documents
Premixed buffer powders that simply require adding water to generate a ready-to-use buffer.
Documents
Specifications
Specifications
  • VPE:
    2
  • Omschrijving:
    Phosphate buffered saline (PBS), JAW™, 10X, dry buffer pack for 10 l of 1X/ pack
  • pH-waarde:
    7,4
  • Bestanddelen:
    43 mM NaH₂PO₄, 1,37 M NaCl, 27mM KCl and 14 mM KH₂PO₄
Specifications