Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Test Strips, EDTA

Test Strips, EDTA

Diaquant® peroxide, 0 • 1 • 3 • 10 • 30 • 100 - mg/L - H₂O₂, 40×40×120 mm (packaging), 100 Tests, 100
Catalogus # MANA932002
Leverancier:   MACHEREY-NAGEL
Test Strips, EDTA
Catalogus # MANA932002
Leverancier:  MACHEREY-NAGEL
CAS-nummer:  
Test Strips, EDTA
Test Strips, EDTA

Diaquant® peroxide, 0 • 1 • 3 • 10 • 30 • 100 - mg/L - H₂O₂, 40×40×120 mm (packaging), 100 Tests, 100

CAS-nummer:  
Price
33,10 €
Log in voor beschikbaarheid
1 * 100 ST
Product Details & Documents

QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. These test strips are immediately ready-to-use. They do not require additional accessories. The test strips are intended for single use, maintenance or calibration are not required.

  • Results within seconds: Just dip, wait and read against multi-stage colour scale Always ready to use: No accessories necessary for evaluation Safe results: Desiccant in the stopper for optimal protection of the strips against humidity

These test strips are for the rapid and reliable determination of EDTA, NTA and other complexing agents in solutions. The easy dip-and-read-procedure provides a reliable result within 15 seconds.

Specificaties

Specificaties

  • VPE:
    100
  • Omschrijving:
    Diaquant® peroxide
  • Afmetingen:
    40×40×120 mm (packaging)
  • Tests per kit:
    100 Tests
  • Meetbereik:
    0 • 1 • 3 • 10 • 30 • 100 - mg/L - H₂O₂
  • Cat. No.:
    MANA932002
  • Platform:
    Semi-quantitative test strips
  • Instrument:
    No
  • Parameter:
    All reagents included
  • Method:
    Peroxide - white → blue
  • Bestanddelen:
    100 test strips in a tube, instruction leaflet
  • Gewicht:
    45 g
  • Conformiteit:
    No hazardous material
  • Houdbaarheid (in maanden):
    30 (from production)

Specificaties