Natriumhypochloriet (11 - 14% Cl₂) in aqueous solution
Glass bottle, 500 mL
Natriumhypochloriet (11 - 14% Cl₂) in aqueous solution
L14709.AP
: 7681-52-9
- :7681-52-9
- :ClNaO
- :MFCD00011120
- :74.44 g/mol
- :111 °C (1013 hPa)
- :–30…–20 °C
- :1,22…1.25 g/cm³ (20 °C)
- :Koelkast
- :231-668-3
- :1791
- :8,II
- Verpakking:Glass bottle
- VPE:500 mL