Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Natriumhypochloriet (11 - 14% Cl₂) in aqueous solution
Glass bottle, 500 mL
  L14709.AP
Natriumhypochloriet (11 - 14% Cl₂) in aqueous solution
  L14709.AP
 :  7681-52-9
Natriumhypochloriet (11 - 14% Cl₂) in aqueous solution
Natriumhypochloriet (11 - 14% Cl₂) in aqueous solution
Glass bottle, 500 mL
 :  7681-52-9
33,60 €
1 * 500 mL
  •  :7681-52-9
  •  :ClNaO
  •  :MFCD00011120
  •  :74.44 g/mol
  •  :111 °C (1013 hPa)
  •  :–30…–20 °C
  •  :1,22…1.25 g/cm³ (20 °C)
  •  :Koelkast
  •  :231-668-3
  •  :1791
  •  :8,II
  • Verpakking:
    Glass bottle
  • VPE:
    500 mL