Bulb pipettes, Qualicolor, colour coded (one mark), Class B
200 ml, 0,200 ± ml, blauw
Catalogus # KAVA632433131037
Leverancier: Kavalier
Bulb pipettes, Qualicolor, colour coded (one mark), Class B
Catalogus # KAVA632433131037
Leverancier: Kavalier
Ga naar:
Product Details & Documents
Soda-lime glass.
- Enlarged form
- Not autoclavable at 121 °C
: DIN 12690, ISO 648
Specificaties
- Kleurcode:blauw
- Capaciteit:200 ml
- Seq:14
- Tolerantie:0,200 ± ml



