Standard for osmometer
400 mOsm calibration standard, 10×2 ml, 10
Leverancier: Advanced Instruments
Standard for osmometer
Catalogus # ADII3MA040
Leverancier: Advanced Instruments
Marketsource
CAS-nummer:
Ga naar:
Product Details & Documents
Calibration standard is used to check the perforance of osmometer and test methods.
Certificaten
Zoeken op lotnummerSpecificaties
Specificaties
- VPE:10
- Omschrijving:400 mOsm calibration standard, 10×2 ml