Basic Violet 3, Sigma-Aldrich®
Glass bottle for solids, 100 g
Leverancier: SIGMA ALDRICH MICROSCOPY
Basic Violet 3, Sigma-Aldrich®
Catalogus # C0775-100G
Leverancier: SIGMA ALDRICH MICROSCOPY
CAS-nummer: 548-62-9
Ga naar:
Product Details & Documents
Synoniemen: Methylrosanilide chloride, Basic violet 3, Viocid, N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanamonium chloride, Violet crystal, Methyl violet 10B, Dimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-ylidene)ammonium chloride, CI 42555, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride, Hexamethyl violet, Hexamethyl-p-rosaniline chloride, Vianin, Gentian violet
- CAS-nummer:548-62-9
- Formule:C₂₅H₃₀ClN₃
- MDL-nummer:MFCD00011750
- Molecular Weight:407.99 g/mol
- Merck index:13,04406
- Smelten Pt:189…194 °C
- Dichtheid:1,19 g/cm³ (20 °C)
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:208-953-6
- UN:3077
- ADR:9,III
Specificaties
Specificaties
- Verpakking:Glass bottle for solids
- VPE:100 g



