Filling cannulae
Filling cannulae, G 16, 76 mm
Catalogus # MURRM716/05
Leverancier: MURRAY S
Ga naar:
Product Details & Documents
Filling cannulae with blunt end and Luer lock fitting.
Bestelinformatie: Different sizes and gauges are available.
Documents
MURRM714^06_TDS_EN
Technische fiche
Specificaties
- Meter:G 16
- Omschrijving:Filling cannulae
- Lengte:76 mm
- Seq:008