Filling cannulae
Filling cannulae, G 14, 100 mm
Catalogus # MURRM714/06
Leverancier: MURRAY S
Ga naar:
Product Details & Documents
Filling cannulae with blunt end and Luer lock fitting.
Ordering information: Different sizes and gauges are available.
Documents
MURRM714^06_TDS_EN
Datasheet
Specificaties
- Meter:G 14
- Omschrijving:Filling cannulae
- Lengte:100 mm
- Seq:012