Hypodermic needles
Hypodermic, sharp, 16, 150 mm
Catalogus # MURRM616/08
Leverancier: MURRAY S
Ga naar:
Product Details & Documents
Non sterile needles with sharp ends and luer lock fitting.
Specificaties
- Meter:16
- Omschrijving:Hypodermic, sharp
- Lengte:150 mm
- Seq:12