Orders LinkContactUsLinkComponent
Hypodermic needles
Hypodermic, sharp, 14, 150 mm
Catalogus # MURRM614/08
Leverancier:   MURRAY S
Hypodermic needles
Catalogus # MURRM614/08
Leverancier:  MURRAY S
Hypodermic needles
Hypodermic needles
Hypodermic, sharp, 14, 150 mm
Price
65,10 €
Log in voor beschikbaarheid
1 * 10 ST
Product Details & Documents
Non sterile needles with sharp ends and luer lock fitting.
Specificaties
  • Meter:
    14
  • Omschrijving:
    Hypodermic, sharp
  • Lengte:
    150 mm
  • Seq:
    08