Hypodermic needles
Hypodermic, sharp, 12, 76 mm
Catalogus # MURRM612/05
Leverancier: MURRAY S
Ga naar:
Product Details & Documents
Non sterile needles with sharp ends and luer lock fitting.
Specificaties
- Meter:12
- Omschrijving:Hypodermic, sharp
- Lengte:76 mm
- Seq:02