Transport container with screw cap
120 ml, 44 mm, 114 mm, with insert/print, 250/250 (bag/ctn)
Catalogus # BOET08.033.0600
Leverancier: BOETTGER PAUL
Ga naar:
Product Details & Documents
Made of PP, translucent. Mailing containers for medical and biohazard samples.
- Optimal protection from external elements through cylindrical design
- Absorbent paper insert provides additional protection, easy to change, multiple-use very simple
- Biohazard and recycling symbol clearly visible
Mailing containers Ø 30 mm fits into current mailboxes with 35 mm-wide slot, containers 44/114 mm suitable for mailing common culture mediums and containers 30/126 mm with cap suitable for Port-a-Cul.
: Corresponding to DIN EN 829 (with insert/print).
: Transport tubes and screw caps have to be ordered separate.
: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904
Specificaties
- Ø uitw.:44 mm
- Verpakt:250/250 (bag/ctn)
- Uitvoering:with insert/print
- Capaciteit:120 ml
- Hoogte:114 mm
- Seq:09



