Ribbons for label printer BBP™11
Thermal transfer printer ribbon, 7950 series, width 85 mm, BBP™11, 70 m, zwart
Catalog # BRDY804468
Supplier: Brady
Jump to:
Product Details & Documents
Documents
Specifications
- Kleur:zwart
- Omschrijving:Thermal transfer printer ribbon, 7950 series, width 85 mm
- Lengte:70 m
- Seq:08
- Voor:BBP™11
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



