Orders LinkContactUsLinkComponent
Biotin kits, Micro HOOK™
Biotin kits, Micro HOOK™
HOOK™-sulfo-NHS-LC-Biotin Kit, 1 KIT
Catalogus # 786-695
Leverancier:   G-Biosciences
Biotin kits, Micro HOOK™
Catalogus # 786-695
Leverancier:  G-Biosciences
CAS-nummer:  
Biotin kits, Micro HOOK™
Biotin kits, Micro HOOK™
HOOK™-sulfo-NHS-LC-Biotin Kit, 1 KIT
CAS-nummer:  
Price
483,00 €
Log in voor beschikbaarheid
1 * 1 KIT
Product Details & Documents
The micro HOOK™ Biotin kits are designed to label small amounts of proteins, with each kit designed for 8 to 10 labellings of 50 to 250 µg protein/reaction.
  • Micro kit for labelling protein primary amines
  • Optimizer Buffer™ for improved coupling efficiency
  • Gel filtration columns for rapid (<10 minute) purification
  • Labels 50 to 250µg protein/reaction
  • Suitable for 8 to 10 couplings
Kits contain the following Biotin reagents
HOOK™ Sulfo-NHS-Biotin - amine reactive reagent, shortest spacer arm
HOOK™ Sulfo-NHS-LC-Biotin - amine reactive reagent, longer spacer arm
HOOK™ Sulfo-NHS-SS-Biotin - cleavable, amine reactive reagent
HOOK™ NHS-dPEG₄-Biotin - amine reactive pegylated reagent
Specificaties
Specificaties
  • VPE:
    1 KIT
  • Omschrijving:
    HOOK™-sulfo-NHS-LC-Biotin Kit
Specificaties
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.