Double bosshead
Double bosshead with M8 stainless steel thumb bolt, 16 mm
Catalog # JUCHVL04810S
Supplier: JUCHHEIM Laborgerate GmbH
Jump to:
Product Details & Documents
Double bosshead made from epoxy coated malleable iron.
Certifications: DIN 12895
Specifications
- Omschrijving:Double bosshead with M8 stainless steel thumb bolt
- Voor staaf Ø:16 mm