Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Natriumhypochloriet (11 - 14% Cl₂) in aqueous solution

Natriumhypochloriet (11 - 14% Cl₂) in aqueous solution

Glass bottle, 2.500 mL
Catalogus # L14709.0F
Corrosief
Milieu
Leverancier:   Thermo Fisher Scientific
Natriumhypochloriet (11 - 14% Cl₂) in aqueous solution
Catalogus # L14709.0F
CAS-nummer:  7681-52-9
Natriumhypochloriet (11 - 14% Cl₂) in aqueous solution
Natriumhypochloriet (11 - 14% Cl₂) in aqueous solution

Glass bottle, 2.500 mL

CAS-nummer:  7681-52-9
Price
74,40 €
Log in voor beschikbaarheid
1 * 2.500 mL
Product Details & Documents
  • CAS-nummer:7681-52-9
  • Formule:ClNaO
  • MDL-nummer:MFCD00011120
  • Molecular Weight:74.44 g/mol
  • Kokend Pt:111 °C (1013 hPa)
  • Smelten Pt:–30…–20 °C
  • Dichtheid:1,22…1.25 g/cm³ (20 °C)
  • Temperatuur bij opslag:Koelkast
  • EINECS:231-668-3
  • UN:1791
  • ADR:8,II
Specificaties

Specificaties

  • Verpakking:
    Glass bottle
  • VPE:
    2.500 mL

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.