Polystyrene standards, GPC/SEC columns calibration
Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,07, quantities 01 g, 1
Catalogus # VARIPL2012-4001
Leverancier: VARIAN
Polystyrene standards, GPC/SEC columns calibration
Catalogus # VARIPL2012-4001
Leverancier: VARIAN
CAS-nummer:
Ga naar:
Product Details & Documents
Polystyrene standards for many organic solvents either for conventional GPC column calibration or for calibrating light scattering and viscosity detectors that provide equidistant calibration points for greater accuracy.
- Compatible with most organic solvents
- Certificate of analysis meets international protocols
- Calibration capability for virtually all applications
Documents
Certificaten
Zoeken op lotnummerSpecificaties
Specificaties
- VPE:1
- Omschrijving:Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,07, quantities 01 g