Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Gas clean filter
Gas clean filter
Gas Clean high-flow connecting unit, 2-position, with 6,35 mm fittings, for high-flow applications such as ICP-MS or ICP-OES
Catalog # VARICP17984
Supplier:   VARIAN
Gas clean filter
Catalog # VARICP17984
Supplier:  VARIAN
Gas clean filter
Gas clean filter
Gas Clean high-flow connecting unit, 2-position, with 6,35 mm fittings, for high-flow applications such as ICP-MS or ICP-OES
Price
666,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Schone gassen verkleinen het risico op kolomschade en vergroten de nauwkeurigheid en gevoeligheid. Deze onbreekbare filters met eigen indicatie bieden een hoge gevoeligheid voor zuurstof en vocht en kunnen in een paar seconden zonder gereedschap worden geïnstalleerd.
Documents
Specifications
  • Omschrijving:
    Gas Clean high-flow connecting unit, 2-position, with 6,35 mm fittings, for high-flow applications such as ICP-MS or ICP-OES
  • Seq:
    21
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.