Orders LinkContactUsLinkComponent
Safety helmet with integrated spectacles
Safety helmet with integrated spectacles
Helmet with integrated spectacles, oranje
Catalogus # SINGCASIRISOR2
Leverancier:   SINGER safety
Safety helmet with integrated spectacles
Catalogus # SINGCASIRISOR2
Leverancier:  SINGER safety
Safety helmet with integrated spectacles
Safety helmet with integrated spectacles
Helmet with integrated spectacles, oranje
Price
71,40 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Polyamide working helmet IRIS2 with clear polycarbonate flip-up spectacles.
  • Adjustable ventilation
  • Ratchet adjustement
Ideal for use in industry, public works professions, maintenance of parks and gardens.
Specificaties
  • Kleur:
    oranje
  • Omschrijving:
    Helmet with integrated spectacles
  • Seq:
    03