Safety helmet with integrated spectacles
Helmet with integrated spectacles, oranje
Catalogus # SINGCASIRISOR2
Leverancier: SINGER safety
Ga naar:
Product Details & Documents
Polyamide working helmet IRIS2 with clear polycarbonate flip-up spectacles.
- Adjustable ventilation
- Ratchet adjustement
Ideal for use in industry, public works professions, maintenance of parks and gardens.
Specificaties
- Kleur:oranje
- Omschrijving:Helmet with integrated spectacles
- Seq:03
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.